missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Tyr-Val-Ala-Asp fluoroacyloxymethyl ketone

For cellular and molecular biology applications

Specifications

Product Type N-Acetyl-Tyr-Val-Ala-Asp fluoroacyloxymethyl ketone
Molecular Weight 762.65
Storage Requirements -30° to -10°C
Sequence ICE Inhibitor III
Format Lyophilized powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66655MCR View Documents Alfa Aesar™
J66655MCR
1mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66655EXD View Documents Alfa Aesar™
J66655EXD
0.5mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

N-Acetyl-Tyr-Val-Ala-Asp fluoroacyloxymethyl ketone
-30° to -10°C
Lyophilized powder
RUO
762.65
ICE Inhibitor III
Ac-YVAD-FAOM
HPLC

For Research Use Only