Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Tyr-(N-Methyl)-Val-Ala-(N-Methyl)-Asp-al

For cellular and molecular biology applications

$73.69 - $107.43

Specifications

Product Type N-Acetyl-Tyr-(N-Methyl)-Val-Ala-(N-Methyl)-Asp-al
Molecular Weight 532.58
Storage Requirements -30° to -10°C
Sequence N-Acetyl-Tyr-(NMe)-Val-Ala-(NMe)-Asp-CHO
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66571EXD

 
alfa aesar™
J66571EXD
0.5mg Each for $73.69

AAJ66571MCR

 
alfa aesar™
J66571MCR
1mg Each for $107.43
Description & Specifications

Specifications

Product Type N-Acetyl-Tyr-(N-Methyl)-Val-Ala-(N-Methyl)-Asp-al
Molecular Weight 532.58
Storage Requirements -30° to -10°C
Sequence N-Acetyl-Tyr-(NMe)-Val-Ala-(NMe)-Asp-CHO
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC