missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Ser-Gln-Asn-Tyr-Pro-Val-Val-NH_2

For cellular and molecular biology applications

$68.62 - $68.62

Specifications

Product Type N-Acetyl-Ser-Gln-Asn-Tyr-Pro-Val-Val-NH_2
Molecular Weight 846.94
Storage Requirements -30° to -10°C
Sequence Ac-Ser-Gln-Asn-Tyr-Pro-Val-Val-NH2
Format Solid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66080MCR
View Documents
Alfa Aesar™
J66080MCR
1mg Each for $68.62
 
Specifications

Specifications

N-Acetyl-Ser-Gln-Asn-Tyr-Pro-Val-Val-NH_2
-30° to -10°C
Solid
HPLC
846.94
Ac-Ser-Gln-Asn-Tyr-Pro-Val-Val-NH2
RUO

For Research Use Only