Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Ser-Asp-Lys-Pro

$68.35 - $68.35

Chemical Identifiers

Chemical Name or Material N-Acetyl-Ser-Asp-Lys-Pro
CAS 127103-11-1
Formula Weight 487.5
MDL Number MFCD00076849
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66430LB0

 
alfa aesar™
J66430LB0
5mg Each for $68.35
Description & Specifications

Specifications

Formula Weight 487.5
Chemical Name or Material N-Acetyl-Ser-Asp-Lys-Pro
CAS 127103-11-1
MDL Number MFCD00076849
Physical Form Solid
Solubility Information Soluble in water at 2mg/mL
Color White to off-white
Quantity 5mg
Storage Note 1 -30° to -10°C

Applications
N-Acetyl-Ser-Asp-Lys-Pro inhibits the entry of pluripotent hemopoietic stem cells into S-phase of the cell cycle and protects against Ara-C lethality in mice. It is a specific substrate for the N-terminal active site of angiotensin converting enzyme, which is responsible for its degradation in vivo.

Solubility
Soluble in water at 2mg/ml

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Safety and Handling

-30° to -10°C