missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Pepstatin

$387.07 - $646.15

Chemical Identifiers

CAS 28575-34-0
Molecular Formula C31H57N5O9
Molecular Weight (g/mol) 643.823
MDL Number MFCD00214071
InChI Key WKYBEGDEGRCZNF-LBTYKNIQSA-N
Synonym acetylpepstatin, acetyl-pepstatin, acetyl pepstatin, ac-val-val-sta-ala-sta, 3s,4s-4-2s-2-3s,4s-4-2s-2-2s-2-acetamido-3-methyl-butanoyl amino-3-methyl-butanoyl amino-3-hydroxy-6-methyl-heptanoyl amino propanoyl amino-3-hydroxy-6-methyl-heptanoic acid, pepstatin ac, pepsidin c, pepsidine c, 5hvp, n-acetyl pepstatin
PubChem CID 5481345
IUPAC Name (3S,4S)-4-[[(2S)-2-[[(3S,4S)-4-[[(2S)-2-[[(2S)-2-acetamido-3-methylbutanoyl]amino]-3-methylbutanoyl]amino]-3-hydroxy-6-methylheptanoyl]amino]propanoyl]amino]-3-hydroxy-6-methylheptanoic acid
SMILES CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66917MB View Documents Alfa Aesar™
J66917MB
25mg Each for $387.07
 
AAJ66917MF View Documents Alfa Aesar™
J66917MF
50mg Each for $646.15
 
Specifications

Chemical Identifiers

28575-34-0
643.823
WKYBEGDEGRCZNF-LBTYKNIQSA-N
5481345
CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)C)O
C31H57N5O9
MFCD00214071
acetylpepstatin, acetyl-pepstatin, acetyl pepstatin, ac-val-val-sta-ala-sta, 3s,4s-4-2s-2-3s,4s-4-2s-2-2s-2-acetamido-3-methyl-butanoyl amino-3-methyl-butanoyl amino-3-hydroxy-6-methyl-heptanoyl amino propanoyl amino-3-hydroxy-6-methyl-heptanoic acid, pepstatin ac, pepsidin c, pepsidine c, 5hvp, n-acetyl pepstatin
(3S,4S)-4-[[(2S)-2-[[(3S,4S)-4-[[(2S)-2-[[(2S)-2-acetamido-3-methylbutanoyl]amino]-3-methylbutanoyl]amino]-3-hydroxy-6-methylheptanoyl]amino]propanoyl]amino]-3-hydroxy-6-methylheptanoic acid

Specifications

White
28575-34-0
MFCD00214071
WKYBEGDEGRCZNF-LBTYKNIQSA-N
(3S,4S)-4-[[(2S)-2-[[(3S,4S)-4-[[(2S)-2-[[(2S)-2-acetamido-3-methylbutanoyl]amino]-3-methylbutanoyl]amino]-3-hydroxy-6-methylheptanoyl]amino]propanoyl]amino]-3-hydroxy-6-methylheptanoic acid
5481345
Solid
25mg
C31H57N5O9
acetylpepstatin, acetyl-pepstatin, acetyl pepstatin, ac-val-val-sta-ala-sta, 3s,4s-4-2s-2-3s,4s-4-2s-2-2s-2-acetamido-3-methyl-butanoyl amino-3-methyl-butanoyl amino-3-hydroxy-6-methyl-heptanoyl amino propanoyl amino-3-hydroxy-6-methyl-heptanoic acid, pepstatin ac, pepsidin c, pepsidine c, 5hvp, n-acetyl pepstatin
CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)C)O
643.823
643.82
N-Acetyl-Pepstatin
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C