Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Leu-Glu-His-Asp-al

Chemical Identifiers

Chemical Name or Material N-Acetyl-Leu-Glu-His-Asp-al
Formula Weight 538.55
MDL Number MFCD01862665
Synonym Acetyl-LEHD-CHO; Caspase-9 Inhibitor
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66787MCR

 
alfa aesar™
J66787MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66787LB0

 
alfa aesar™
J66787LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 538.55
Chemical Name or Material N-Acetyl-Leu-Glu-His-Asp-al
Synonym Acetyl-LEHD-CHO; Caspase-9 Inhibitor
Physical Form Lyophilized Powder
Color White to off-white
Storage Note 1 -30° to -10°C
Sensitivity Light Sensitive
Quantity 1mg
MDL Number MFCD01862665
Safety and Handling

-30° to -10°C