Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Leu-Glu-His-Asp-al


Quantity 1mg

Manufacturer: alfa aesar™  J66787MCR

 View more versions of this product

Catalog No. AAJ66787MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 538.55
Chemical Name or Material N-Acetyl-Leu-Glu-His-Asp-al
Synonym Acetyl-LEHD-CHO; Caspase-9 Inhibitor
Physical Form Lyophilized Powder
Color White to off-white
Storage Note 1 -30° to -10°C
Sensitivity Light Sensitive
Quantity 1mg
MDL Number MFCD01862665

Specifications

Chemical Name or Material N-Acetyl-Leu-Glu-His-Asp-al
Formula Weight 538.55
MDL Number MFCD01862665
Synonym Acetyl-LEHD-CHO; Caspase-9 Inhibitor
Accessories

-30° to -10°C