missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-L-Tyrosine, MP Biomedicals™

Manufacturer:  Mp Biomedicals 10015005

 View more versions of this product

Catalog No. ICN10015005



Specifications

Specifications

N-Acetyl-L-Tyrosine
C11H13NO4
CAHKINHBCWCHCF-JTQLQIEISA-N
(2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
68310
223.2
Solid
537-55-3
ac-tyr-oh
CC(=O)NC(CC1=CC=C(C=C1)O)C(=O)O
223.228
CHEBI:21563
Crystalline
5g

Chemical Identifiers

537-55-3
223.228
ac-tyr-oh
CHEBI:21563
CC(=O)NC(CC1=CC=C(C=C1)O)C(=O)O
C11H13NO4
CAHKINHBCWCHCF-JTQLQIEISA-N
68310
(2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
Safety and Handling

Safety and Handling

EINECSNumber : 208-671-3

Recommended Storage : Store at 0°C.

Documents