Please login to your online account to display your discounted pricing

N-Acetyl-L-tyrosine, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  347020250

 View more versions of this product

Catalog No. AC347020250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Ammonium 200ppm max.
Arsenic (As) 1ppm max.
Chlorine (Cl) 300ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Loss on Drying 0.3% max.
Packaging Glass bottle
Specific Rotation +45° to +50° (20°C, 589nm) (c=2, water)
Sulfated Ash 0.1% max.
Sulfate 300ppm max.
Transmittance 98% min. (c=5, H2O, 430nm) 1cm cell
Color White
Quantity 25g
Chemical Name or Material N-Acetyl-L-tyrosine
Name Note 98%
CAS 537-55-3
CAS Min % 97.5
CAS Max % 100.0
Physical Form White crystalline powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C11H13NO4
Linear Formula OHC6H4CH2CH(NHCOCH3)COOH
Formula Weight 223.23g/mol
MDL Number MFCD00037190
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 208-671-3
TSCA TSCA

Specifications

Chemical Name or Material N-Acetyl-L-tyrosine
Name Note 98%
CAS 537-55-3
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C11H13NO4
Linear Formula OHC6H4CH2CH(NHCOCH3)COOH
Formula Weight 223.23g/mol
MDL Number MFCD00037190
Accessories

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.

SDS & Safety Documents