missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Acetyl-L-tryptophan methyl ester, 95%, Thermo Scientific Chemicals
$133.36 - $480.70
Chemical Identifiers
CAS | 2824-57-9 |
---|---|
Molecular Formula | C14H16N2O3 |
Molecular Weight (g/mol) | 260.29 |
MDL Number | MFCD00022759 |
InChI Key | XZECNVJPYDPBAM-ZDUSSCGKSA-N |
Synonym | methyl n-acetyl-l-tryptophanate, ac-trp-ome, methyl acetyltryptophanate, l-tryptophan, n-acetyl-, methyl ester, unii-mup4y18zq5, n-acetyltryptophan methyl ester, n-acetyl-l-tryptophan methyl ester, mup4y18zq5, methyl 2s-2-acetamido-3-1h-indol-3-yl propanoate, l-tryptophan,n-acetyl-, methyl ester |
PubChem CID | 102337 |
IUPAC Name | methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate |
SMILES | COC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6693706
|
Thermo Scientific Chemicals
H6693706 |
5 g |
Each for $133.36
|
|
|||||
AAH6693714
|
Thermo Scientific Chemicals
H6693714 |
25 g |
Each for $480.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2824-57-9 | |
260.29 | |
XZECNVJPYDPBAM-ZDUSSCGKSA-N | |
102337 | |
COC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(C)=O |
C14H16N2O3 | |
MFCD00022759 | |
methyl n-acetyl-l-tryptophanate, ac-trp-ome, methyl acetyltryptophanate, l-tryptophan, n-acetyl-, methyl ester, unii-mup4y18zq5, n-acetyltryptophan methyl ester, n-acetyl-l-tryptophan methyl ester, mup4y18zq5, methyl 2s-2-acetamido-3-1h-indol-3-yl propanoate, l-tryptophan,n-acetyl-, methyl ester | |
methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate |
Specifications
2824-57-9 | |
C14H16N2O3 | |
methyl n-acetyl-l-tryptophanate, ac-trp-ome, methyl acetyltryptophanate, l-tryptophan, n-acetyl-, methyl ester, unii-mup4y18zq5, n-acetyltryptophan methyl ester, n-acetyl-l-tryptophan methyl ester, mup4y18zq5, methyl 2s-2-acetamido-3-1h-indol-3-yl propanoate, l-tryptophan,n-acetyl-, methyl ester | |
COC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(C)=O | |
260.29 | |
260.29 | |
5 g | |
N-Acetyl-L-tryptophan methyl ester |
White | |
MFCD00022759 | |
XZECNVJPYDPBAM-ZDUSSCGKSA-N | |
methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate | |
102337 | |
95% | |
Powder |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only