Please login to your online account to display your discounted pricing

N-acetyl-l-proline, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC376640050

 View more versions of this product

Catalog No. AC376640050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥98.5%
Formula Weight 157.17
Quantity 5g
Color White to off-white
Specific Rotation Condition −86° (20°C c=1 EtOH)
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C7H11NO3
CAS Min % 98.5
Chemical Name or Material N-Acetyl-L-proline
Packaging Glass bottle
MDL Number MFCD00020837
CAS 68-95-1
Specific Rotation -84° to -88° (20°C, 589nm) (c=1, EtOH)

Specifications

Chemical Name or Material N-Acetyl-L-proline
CAS 68-95-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H11NO3
Formula Weight 157.17
MDL Number MFCD00020837