missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Acetyl-L-phenylalanyl-3,5-diiodo-L-tyrosine, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 227652500
Specifications
N-Acetyl-L-phenylalanyl-3, 5-diiodo-L-tyrosine | |
Glass bottle | |
219.0°C to 221.0°C | |
95% | |
94.0 | |
94% min | |
MFCD00002180 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,and alkalies | |
CC(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC1=CC(I)=C(O)C(I)=C1)C(O)=O | |
622.2 | |
622.2 | |
95% |
Authentic | |
White | |
250mg | |
3786-08-1 | |
100.0 | |
C20H20I2N2O5 | |
anforstan, berthollet salt, berthollet's salt, chlorate of potash, chloric acid, potassium salt, fekabit, kaliumchlorat, potassium chlorate, potcrate, salt of tarter | |
DWIHCEZUWCRUES-UHFFFAOYNA-N | |
potassium;chlorate | |
6426889 | |
Crystals or Crystalline Powder |
Chemical Identifiers
3786-08-1 | |
622.2 | |
DWIHCEZUWCRUES-UHFFFAOYNA-N | |
6426889 | |
CC(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC1=CC(I)=C(O)C(I)=C1)C(O)=O |
C20H20I2N2O5 | |
MFCD00002180 | |
anforstan, berthollet salt, berthollet's salt, chlorate of potash, chloric acid, potassium salt, fekabit, kaliumchlorat, potassium chlorate, potcrate, salt of tarter | |
potassium;chlorate |
Safety and Handling
EINECSNumber : 223-254-6