missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-L-phenylalanyl-3,5-diiodo-L-tyrosine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 227650500

 View more versions of this product

Catalog No. AC227650500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
622.2
White
100.0
95%
95%
3786-08-1
Authentic
Crystals or Crystalline Powder
50mg
94.0
Glass bottle
C20H20I2N2O5
MFCD00002180
N-Acetyl-L-phenylalanyl-3, 5-diiodo-L-tyrosine

Chemical Identifiers

3786-08-1
100.0
C20H20I2N2O5
MFCD00002180
≥94%
94.0
95%
622.2
N-Acetyl-L-phenylalanyl-3, 5-diiodo-L-tyrosine, 95%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 223-254-6