Please login to your online account to display your discounted pricing

N-Acetyl-L-phenylalanyl-3,5-diiodo-L-tyrosine, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Acetyl-L-phenylalanyl-3, 5-diiodo-L-tyrosine
Name Note 95%
CAS 3786-08-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C20H20I2N2O5
Formula Weight 622.2
MDL Number MFCD00002180
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC227652500

  SDS
thermo fisher scientific
227652500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC227650500

  SDS
thermo fisher scientific
227650500
Glass bottle 50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystals or Crystalline Powder
Formula Weight 622.2
Quantity 250mg
Color White
EINECS Number 223-254-6
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material N-Acetyl-L-phenylalanyl-3, 5-diiodo-L-tyrosine
Infrared Spectrum Authentic
Name Note 95%
Assay Percent Range 95%
Molecular Formula C20H20I2N2O5
MDL Number MFCD00002180
CAS 3786-08-1
Packaging Glass bottle