Please login to your online account to display your discounted pricing

N-Acetyl-L-phenylalanine, 99+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  227930010

 View more versions of this product

Catalog No. AC227930010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Loss on Drying 1% max. (105°C, 4 hrs)
Packaging Glass bottle
Specific Rotation +39° to +41° (20°C, 589nm) (c=1, MeOH)
Water 0.5% max. (K.F.)
Color White to off-white
Quantity 1g
Chemical Name or Material N-Acetyl-L-phenylalanine
Name Note 99+%
CAS 2018-61-3
CAS Min % 99.0
CAS Max % 100.0
Physical Form White to off-white fine crystalline powder or needles
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C11H13NO3
Linear Formula C6H5CH2CH(NHCOCH3)CO2H
Formula Weight 207.23g/mol
MDL Number MFCD00063158
EINECS Number 217-959-8
TSCA TSCA

Specifications

Chemical Name or Material N-Acetyl-L-phenylalanine
Name Note 99+%
CAS 2018-61-3
CAS Min % 99.0
CAS Max % 100.0
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C11H13NO3
Linear Formula C6H5CH2CH(NHCOCH3)CO2H
Formula Weight 207.23g/mol
MDL Number MFCD00063158