Please login to your online account to display your discounted pricing

N-Acetyl-L-phenylalanine, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Acetyl-L-phenylalanine
Name Note 99+%
CAS 2018-61-3
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C11H13NO3
Linear Formula C6H5CH2CH(NHCOCH3)CO2H
Formula Weight 207.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC227930050

  SDS
thermo fisher scientific
227930050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Fine Crystalline Powder or Needles
Formula Weight 207.23
Quantity 5g
Color White to off-white
EINECS Number 217-959-8
TSCA TSCA
CAS Min % 99.0
CAS Max % 100.0
Name Note 99+%
Loss on Drying 1% max. (105°C, 4 hrs)
Linear Formula C6H5CH2CH(NHCOCH3)CO2H
MDL Number MFCD00063158
Molecular Formula C11H13NO3
CAS 2018-61-3
Chemical Name or Material N-Acetyl-L-phenylalanine
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Specific Rotation +39° to +41° (20°C, 589nm) (c=1, MeOH)
Water 0.5% max. (K.F.)