Please login to your online account to display your discounted pricing

N-Acetyl-L-leucyl-L-leucyl-L-methional, Tocris


Cathepsin inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS038410

 View more versions of this product

Catalog No. 038410

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material N-Acetyl-L-leucyl-L-leucyl-L-methional
CAS 136632-32-1
Recommended Storage Store at -20°C
Assay Percent Range >98%
Molecular Formula C19H35N3O4S
Formula Weight 401.56
Quantity 10mg

Specifications

Chemical Name or Material N-Acetyl-L-leucyl-L-leucyl-L-methional
Assay Percent Range >98%
Molecular Formula C19H35N3O4S
Formula Weight 401.56
Accessories

Store at -20°C