Please login to your online account to display your discounted pricing

N-Acetyl-L-leucyl-L-leucyl-L-methional, Tocris

Cathepsin inhibitor

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material N-Acetyl-L-leucyl-L-leucyl-L-methional
Assay Percent Range >98%
Molecular Formula C19H35N3O4S
Formula Weight 401.56
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

038450

 
r & d systems
RDS038450
50mg Each for $697.40

038410

 
r & d systems
RDS038410
10mg Each for $168.15
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 401.56
Chemical Name or Material N-Acetyl-L-leucyl-L-leucyl-L-methional
Assay Percent Range >98%
Molecular Formula C19H35N3O4S
CAS 136632-32-1
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C