missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals N-Acetyl-L-leucine, 99%
$60.53 - $60.53
Chemical Identifiers
CAS | 1188-21-2 |
---|---|
Molecular Formula | C8H15NO3 |
Molecular Weight (g/mol) | 173.21 |
MDL Number | MFCD00065131 |
InChI Key | WXNXCEHXYPACJF-JLDDOWRYNA-N |
Synonym | n-acetyl-l-leucine, acetyl-l-leucine, ac-leu-oh, n-acetylleucine, s-2-acetamido-4-methylpentanoic acid, n-acetyl-leu, 2s-2-acetamido-4-methylpentanoic acid, n-acetyl-leucine, leucine, n-acetyl-, l, n-acetyl-l-leucin |
PubChem CID | 70912 |
ChEBI | CHEBI:17786 |
IUPAC Name | (2S)-2-acetamido-4-methylpentanoic acid |
SMILES | CC(C)C[C@H](NC(C)=O)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC347010050
|
Thermo Scientific Chemicals
347010050 |
5 g | Glass bottle |
Each for $60.53
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1188-21-2 | |
173.21 | |
WXNXCEHXYPACJF-JLDDOWRYNA-N | |
70912 | |
(2S)-2-acetamido-4-methylpentanoic acid |
C8H15NO3 | |
MFCD00065131 | |
n-acetyl-l-leucine, acetyl-l-leucine, ac-leu-oh, n-acetylleucine, s-2-acetamido-4-methylpentanoic acid, n-acetyl-leu, 2s-2-acetamido-4-methylpentanoic acid, n-acetyl-leucine, leucine, n-acetyl-, l, n-acetyl-l-leucin | |
CHEBI:17786 | |
CC(C)C[C@H](NC(C)=O)C(O)=O |
Specifications
1188-21-2 | |
100.0 | |
White | |
99% | |
C8H15NO3 | |
MFCD00065131 | |
− 24.50 | |
(5% in ethanol) Clear colorless | |
CC(C)C[C@H](NC(C)=O)C(O)=O | |
173.21 | |
CHEBI:17786 | |
99% | |
N-Acetyl-L-leucine, 99% |
98.5 | |
190.0°C | |
Authentic | |
Glass bottle | |
(CH3)2CHCH2CH(NHCOCH3)COOH | |
5 g | |
n-acetyl-l-leucine, acetyl-l-leucine, ac-leu-oh, n-acetylleucine, s-2-acetamido-4-methylpentanoic acid, n-acetyl-leu, 2s-2-acetamido-4-methylpentanoic acid, n-acetyl-leucine, leucine, n-acetyl-, l, n-acetyl-l-leucin | |
WXNXCEHXYPACJF-JLDDOWRYNA-N | |
(2S)-2-acetamido-4-methylpentanoic acid | |
70912 | |
173.21 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : 214-706-3
TSCA : TSCA
RUO – Research Use Only