Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-L-leucine, 99%

$50.34 - $164.70

Chemical Identifiers

Chemical Name or Material N-Acetyl-L-leucine
CAS 1188-21-2
Molecular Formula C8H15NO3
Formula Weight 173.21
MDL Number MFCD00065131
Beilstein 1724849
Synonym Ac-Leu-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1392606

 
alfa aesar™
L1392606
5g Each for $50.34

AAL1392614

 
alfa aesar™
L1392614
25g Each for $164.70
Description & Specifications

Specifications

Formula Weight 173.21
Chemical Name or Material N-Acetyl-L-leucine
Assay 99%
Molecular Formula C8H15NO3
MDL Number MFCD00065131
CAS 1188-21-2
TSCA TSCA
Beilstein 1724849
Synonym Ac-Leu-OH
Melting Point 177° to 179°C
Quantity 5g
Optical Rotation −23° (c=5 in Ethanol)