Please login to your online account to display your discounted pricing

N-acetyl-L-histidine, 100.3%, MP Biomedicals™


Quantity 1g

Manufacturer: mp biomedicals  10009201

 View more versions of this product

Catalog No. ICN10009201

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 197.2
Color White
Quantity 1g
Chemical Name or Material N-acetyl-L-histidine
Assay Percent Range 1.003
Molecular Formula C8H11N3O3
CAS 39145-52-3

Specifications

Chemical Name or Material N-acetyl-L-histidine
CAS 39145-52-3
Physical Form Powder
Assay Percent Range 1.003
Molecular Formula C8H11N3O3
Formula Weight 197.2