Please login to your online account to display your discounted pricing

N-acetyl-L-histidine, 100.3%, MP Biomedicals™

$66.63 - $66.63

Chemical Identifiers

Chemical Name or Material N-acetyl-L-histidine
CAS 39145-52-3
Physical Form Powder
Assay Percent Range 1.003
Molecular Formula C8H11N3O3
Formula Weight 197.2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10009201

 
mp biomedicals
10009201
1g Each for $66.63
Description & Specifications

Specifications

Formula Weight 197.2
Physical Form Powder
Color White
Quantity 1g
Chemical Name or Material N-acetyl-L-histidine
Molecular Formula C8H11N3O3
CAS 39145-52-3
Assay Percent Range 1.003