Please login to your online account to display your discounted pricing

N-Acetyl-L-Glutamine, MP Biomedicals™


Quantity 250g

Manufacturer: mp biomedicals  10009983

 View more versions of this product

Catalog No. ICN10009983

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 188.2
Quantity 250g
EINECS Number 219-647-7
Physical Form Crystalline
Physical State Solid
Synonym Aceglutamide
Chemical Name or Material N-Acetyl-L-Glutamine
Molecular Formula C7H12N2O4
MDL Number MFCD00038159
CAS 2490-97-3
Recommended Storage Store at 2° to 8°C.

Specifications

Chemical Name or Material N-Acetyl-L-Glutamine
CAS 2490-97-3
Physical Form Crystalline
Molecular Formula C7H12N2O4
Formula Weight 188.2
MDL Number MFCD00038159
Synonym Aceglutamide
Accessories

Store at 2° to 8°C.