Please login to your online account to display your discounted pricing

N-Acetyl-L-Glutamine, MP Biomedicals™


Quantity 100g

Manufacturer: mp biomedicals  0210009980

 View more versions of this product

Catalog No. ICN10009980

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical State Solid
Quantity 100g
Chemical Name or Material N-Acetyl-L-Glutamine
CAS 2490-97-3
Physical Form Crystalline
Molecular Formula C7H12N2O4
Formula Weight 188.2g/mol
MDL Number MFCD00038159
Synonym Aceglutamide
Recommended Storage Store at 2° to 8°C.
EINECS Number 219-647-7

Specifications

Chemical Name or Material N-Acetyl-L-Glutamine
CAS 2490-97-3
Physical Form Crystalline
Molecular Formula C7H12N2O4
Formula Weight 188.2g/mol
MDL Number MFCD00038159
Synonym Aceglutamide
Accessories

Store at 2° to 8°C.