Please login to your online account to display your discounted pricing

N-Acetyl-L-Glutamine, MP Biomedicals™

$271.74 - $271.74

Chemical Identifiers

Chemical Name or Material N-Acetyl-L-Glutamine
CAS 2490-97-3
Physical Form Crystalline
Molecular Formula C7H12N2O4
Formula Weight 188.2
MDL Number MFCD00038159
Synonym Aceglutamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10009983

 
mp biomedicals
10009983
250g Each for $271.74
Description & Specifications

Specifications

Chemical Name or Material N-Acetyl-L-Glutamine
CAS 2490-97-3
Recommended Storage Store at 2° to 8°C.
EINECS Number 219-647-7
Physical Form Crystalline
Molecular Formula C7H12N2O4
Formula Weight 188.2
MDL Number MFCD00038159
Synonym Aceglutamide
Physical State Solid
Quantity 250g