missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-L-glutamine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC451640250

 View more versions of this product

Catalog No. AC451640250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

N-Acetyl-L-glutamine
0.2% max
25g
97%
aceglutamide, acetyl-l-glutamine, acetylglutamine, acutil-s, l-glutamine, n-acetyl, n-acetyl-l-glutamine, n-acetylglutamine, n-alpha-acetyl-l-glutamine, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid
KSMRODHGGIIXDV-YFKPBYRVSA-N
(2S)-2-acetamido-5-amino-5-oxopentanoic acid
182230
188.18
Glass bottle
206°C
35305-74-9
C7H12N2O4
Solubility in water: soluble
CC(=O)NC(CCC(=O)N)C(=O)O
188.18
CHEBI:21553
97%

Chemical Identifiers

35305-74-9
188.18
aceglutamide, acetyl-l-glutamine, acetylglutamine, acutil-s, l-glutamine, n-acetyl, n-acetyl-l-glutamine, n-acetylglutamine, n-alpha-acetyl-l-glutamine, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid
CHEBI:21553
CC(=O)NC(CCC(=O)N)C(=O)O
C7H12N2O4
KSMRODHGGIIXDV-YFKPBYRVSA-N
182230
(2S)-2-acetamido-5-amino-5-oxopentanoic acid
SDS Documents