missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Acetyl-L-glutamic acid 5-tert-butyl ester, 95%, Thermo Scientific Chemicals
$103.19 - $395.48
Chemical Identifiers
CAS | 84192-88-1 |
---|---|
Molecular Formula | C11H19NO5 |
Molecular Weight (g/mol) | 245.28 |
MDL Number | MFCD00236754 |
InChI Key | FALCCLKESWGSNI-UHFFFAOYNA-N |
Synonym | ac-glu otbu-oh, s-2-acetamido-5-tert-butoxy-5-oxopentanoic acid, 2s-5-tert-butoxy-2-acetamido-5-oxopentanoic acid |
PubChem CID | 7019606 |
SMILES | CC(=O)NC(CCC(=O)OC(C)(C)C)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6387103
|
Thermo Scientific Chemicals
H6387103 |
1 g |
Each for $103.19
|
|
|||||
AAH6387106
|
Thermo Scientific Chemicals
H6387106 |
5 g |
Each for $395.48
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
84192-88-1 | |
245.28 | |
FALCCLKESWGSNI-UHFFFAOYNA-N | |
7019606 |
C11H19NO5 | |
MFCD00236754 | |
ac-glu otbu-oh, s-2-acetamido-5-tert-butoxy-5-oxopentanoic acid, 2s-5-tert-butoxy-2-acetamido-5-oxopentanoic acid | |
CC(=O)NC(CCC(=O)OC(C)(C)C)C(O)=O |
Specifications
84192-88-1 | |
C11H19NO5 | |
ac-glu otbu-oh, s-2-acetamido-5-tert-butoxy-5-oxopentanoic acid, 2s-5-tert-butoxy-2-acetamido-5-oxopentanoic acid | |
CC(=O)NC(CCC(=O)OC(C)(C)C)C(O)=O | |
245.28 | |
245.27 | |
1 g | |
N-Acetyl-L-glutamic acid 5-tert-butyl ester |
White | |
MFCD00236754 | |
FALCCLKESWGSNI-UHFFFAOYNA-N | |
5-(tert-butoxy)-2-acetamido-5-oxopentanoic acid | |
7019606 | |
95% | |
Powder |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only