missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Acetyl-L-glutamic acid 5-tert-butyl ester, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6387106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Acetyl-L-glutamic acid 5-tert-butyl ester | |
White | |
MFCD00236754 | |
FALCCLKESWGSNI-UHFFFAOYNA-N | |
(2S)-2-acetamido-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid | |
7019606 | |
95% | |
Powder |
84192-88-1 | |
C11H19NO5 | |
ac-glu otbu-oh, s-2-acetamido-5-tert-butoxy-5-oxopentanoic acid, 2s-5-tert-butoxy-2-acetamido-5-oxopentanoic acid | |
CC(=O)NC(CCC(=O)OC(C)(C)C)C(O)=O | |
245.28 | |
245.27 | |
5 g |
Chemical Identifiers
84192-88-1 | |
245.28 | |
FALCCLKESWGSNI-UHFFFAOYNA-N | |
7019606 |
C11H19NO5 | |
MFCD00236754 | |
ac-glu otbu-oh, s-2-acetamido-5-tert-butoxy-5-oxopentanoic acid, 2s-5-tert-butoxy-2-acetamido-5-oxopentanoic acid | |
CC(=O)NC(CCC(=O)OC(C)(C)C)C(O)=O |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only