Please login to your online account to display your discounted pricing

N-Acetyl-L-cysteine, 98+%, Alfa Aesar™

$575.58 - $575.58

Chemical Identifiers

Chemical Name or Material N-Acetyl-L-cysteine
CAS 616-91-1
Molecular Formula C5H9NO3S
Formula Weight 163.2
MDL Number MFCD00004880
Beilstein 1724426
Merck Index 14,88
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1540936

  SDS
alfa aesar
A1540936
Each for $575.58
Description & Specifications

Specifications

Formula Weight 163.2
Quantity 500g
Chemical Name or Material N-Acetyl-L-cysteine
TSCA TSCA
Assay ≥98%
Molecular Formula C5H9NO3S
MDL Number MFCD00004880
CAS 616-91-1
Beilstein 1724426
Merck Index 14,88
Melting Point 108° to 111°C
Optical Rotation +5.3° (c=2 in Water)
RTECS Number HA1660000

Solubility
Soluble in water, ethanol, methanol, dimethyl sulfoxide, hot isopropyl alcohol, methyl acetate and ethyl acetate. Insoluble in chloroform and ether.

Notes
Store in a cool place. Incompatible with heavy metals, heavy metal salts, rubber, oxygen and strong oxidizing agents.