missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-L-aspartic acid 4-tert-butyl ester, 95%

$89.30 - $1,060.97

Chemical Identifiers

CAS 117833-18-8
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.248
MDL Number MFCD00236746
InChI Key NKNPTCBHHPHSEA-ZETCQYMHSA-N
Synonym ac-asp otbu-oh, s-2-acetamido-4-tert-butoxy-4-oxobutanoic acid, n-acetyl-l-aspartic acid 4-tert-butyl ester, acetyl-l-aspartic acid beta-tert-butyl ester, 2s-4-tert-butoxy-2-acetamido-4-oxobutanoic acid, ac-asp-otbu, s-3-acetamido-4-tert-butoxy-4-oxobutanoic acid, l-aspartic acid,n-acetyl-,4-1,1-dimethylethyl ester, l-aspartic acid, n-acetyl-, 4-1,1-dimethylethyl ester
PubChem CID 7019596
IUPAC Name (2S)-2-acetamido-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(=O)NC(CC(=O)OC(C)(C)C)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63933MD View Documents Alfa Aesar™
H63933MD
250mg Each for $89.30
 
AAH6393303 View Documents Alfa Aesar™
H6393303
1g Each for $265.24
 
AAH6393306 View Documents Alfa Aesar™
H6393306
5g Each for $1,060.97
 
Specifications

Chemical Identifiers

117833-18-8
231.248
NKNPTCBHHPHSEA-ZETCQYMHSA-N
7019596
CC(=O)NC(CC(=O)OC(C)(C)C)C(=O)O
C10H17NO5
MFCD00236746
ac-asp otbu-oh, s-2-acetamido-4-tert-butoxy-4-oxobutanoic acid, n-acetyl-l-aspartic acid 4-tert-butyl ester, acetyl-l-aspartic acid beta-tert-butyl ester, 2s-4-tert-butoxy-2-acetamido-4-oxobutanoic acid, ac-asp-otbu, s-3-acetamido-4-tert-butoxy-4-oxobutanoic acid, l-aspartic acid,n-acetyl-,4-1,1-dimethylethyl ester, l-aspartic acid, n-acetyl-, 4-1,1-dimethylethyl ester
(2S)-2-acetamido-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid

Specifications

117833-18-8
MFCD00236746
NKNPTCBHHPHSEA-ZETCQYMHSA-N
(2S)-2-acetamido-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
7019596
Solid
250mg
C10H17NO5
ac-asp otbu-oh, s-2-acetamido-4-tert-butoxy-4-oxobutanoic acid, n-acetyl-l-aspartic acid 4-tert-butyl ester, acetyl-l-aspartic acid beta-tert-butyl ester, 2s-4-tert-butoxy-2-acetamido-4-oxobutanoic acid, ac-asp-otbu, s-3-acetamido-4-tert-butoxy-4-oxobutanoic acid, l-aspartic acid,n-acetyl-,4-1,1-dimethylethyl ester, l-aspartic acid, n-acetyl-, 4-1,1-dimethylethyl ester
CC(=O)NC(CC(=O)OC(C)(C)C)C(=O)O
231.248
231.25
95%
N-Acetyl-L-aspartic acid 4-tert-butyl ester
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C