missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-L-aspartic acid 4-tert-butyl ester, 95%

Manufacturer:  Alfa Aesar™ H6393306

 View more versions of this product

Catalog No. AAH6393306



Specifications

Specifications

N-Acetyl-L-aspartic acid 4-tert-butyl ester
C10H17NO5
ac-asp otbu-oh, s-2-acetamido-4-tert-butoxy-4-oxobutanoic acid, n-acetyl-l-aspartic acid 4-tert-butyl ester, acetyl-l-aspartic acid beta-tert-butyl ester, 2s-4-tert-butoxy-2-acetamido-4-oxobutanoic acid, ac-asp-otbu, s-3-acetamido-4-tert-butoxy-4-oxobutanoic acid, l-aspartic acid,n-acetyl-,4-1,1-dimethylethyl ester, l-aspartic acid, n-acetyl-, 4-1,1-dimethylethyl ester
CC(=O)NC(CC(=O)OC(C)(C)C)C(=O)O
231.248
231.25
95%
117833-18-8
MFCD00236746
NKNPTCBHHPHSEA-ZETCQYMHSA-N
(2S)-2-acetamido-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
7019596
Solid
5g

Chemical Identifiers

117833-18-8
231.248
NKNPTCBHHPHSEA-ZETCQYMHSA-N
7019596
CC(=O)NC(CC(=O)OC(C)(C)C)C(=O)O
C10H17NO5
MFCD00236746
ac-asp otbu-oh, s-2-acetamido-4-tert-butoxy-4-oxobutanoic acid, n-acetyl-l-aspartic acid 4-tert-butyl ester, acetyl-l-aspartic acid beta-tert-butyl ester, 2s-4-tert-butoxy-2-acetamido-4-oxobutanoic acid, ac-asp-otbu, s-3-acetamido-4-tert-butoxy-4-oxobutanoic acid, l-aspartic acid,n-acetyl-,4-1,1-dimethylethyl ester, l-aspartic acid, n-acetyl-, 4-1,1-dimethylethyl ester
(2S)-2-acetamido-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents