Please login to your online account to display your discounted pricing

N-acetyl-L-arginine dihydrate, >99%, MP Biomedicals™

$292.41 - $292.41

Chemical Identifiers

Chemical Name or Material N-acetyl-L-arginine dihydrate
CAS 155-84-0
Physical Form Powder
Assay Percent Range >99%
Molecular Formula C8H16N4O3·2H2O
Formula Weight 252.27
MDL Number MFCD00019733
Synonym 2-Acetamido-5-Guanidinovaleric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10007810

 
mp biomedicals
10007810
10g Each for $292.41
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 252.27
Color White
Quantity 10g
EINECS Number 205-846-6
Assay Percent Range >99%
Molecular Formula C8H16N4O3·2H2O
MDL Number MFCD00019733
CAS 155-84-0
Synonym 2-Acetamido-5-Guanidinovaleric acid
Chemical Name or Material N-acetyl-L-arginine dihydrate