Please login to your online account to display your discounted pricing

N-Acetyl-L-alanyl-L-alanyl-L-alanine, 98%, ACROS Organics™


Quantity 100mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  227611000

 View more versions of this product

Catalog No. AC227611000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 273.28
Quantity 100mg
Color White
EINECS Number 242-912-3
CAS Max % 100.0
Chemical Name or Material N-Acetyl-L-alanyl-L-alanyl-L-alanine
Assay Percent Range 98%
Linear Formula CH3CONH[CH(CH3)CONH]2CH(CH3)CO2H
Infrared Spectrum Authentic
Molecular Formula C11H19N3O5
MDL Number MFCD00002642
CAS 19245-85-3
CAS Min % 97.5
Packaging Glass bottle
Elemental Analysis 97.5% min.

Specifications

Chemical Name or Material N-Acetyl-L-alanyl-L-alanyl-L-alanine
CAS 19245-85-3
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C11H19N3O5
Linear Formula CH3CONH[CH(CH3)CONH]2CH(CH3)CO2H
Formula Weight 273.28
MDL Number MFCD00002642