Please login to your online account to display your discounted pricing

N-Acetyl-L-alanyl-L-alanyl-L-alanine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Acetyl-L-alanyl-L-alanyl-L-alanine
CAS 19245-85-3
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C11H19N3O5
Linear Formula CH3CONH[CH(CH3)CONH]2CH(CH3)CO2H
Formula Weight 273.28
MDL Number MFCD00002642
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC227610250

  SDS
thermo fisher scientific
227610250
Glass bottle 25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC227611000

  SDS
thermo fisher scientific
227611000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 273.28
Physical Form Powder
Quantity 25mg
Color White
EINECS Number 242-912-3
CAS Max % 100.0
Molecular Formula C11H19N3O5
Linear Formula CH3CONH[CH(CH3)CONH]2CH(CH3)CO2H
Assay Percent Range 98%
MDL Number MFCD00002642
CAS Min % 97.5
Chemical Name or Material N-Acetyl-L-alanyl-L-alanyl-L-alanine
CAS 19245-85-3
Infrared Spectrum Authentic
Elemental Analysis 97.5% min.
Packaging Glass bottle