Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-L-alanine methyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Acetyl-L-alanine methyl ester
CAS 3619-02-1
Formula Weight 145.16
MDL Number MFCD03940666
Beilstein 1723226
Synonym Ac-Ala-OMe
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6699006

 
alfa aesar™
H6699006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6699014

 
alfa aesar™
H6699014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 145.16
Chemical Name or Material N-Acetyl-L-alanine methyl ester
MDL Number MFCD03940666
CAS 3619-02-1
Assay 95%
Storage Note 1 -30° to -10°C
Color Yellow
Synonym Ac-Ala-OMe
Beilstein 1723226
Quantity 5g
Physical Form Liquid
Safety and Handling

-30° to -10°C