Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Ile-Glu-Thr-Asp-al

$103.22 - $307.38

Chemical Identifiers

Chemical Name or Material N-Acetyl-Ile-Glu-Thr-Asp-al
CAS 191338-86-0
Formula Weight 502.51
MDL Number MFCD01318858
Synonym Acetyl-IETD-CHO; Caspase-8 Inhibitor I
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66985LB0

 
alfa aesar™
J66985LB0
5mg Each for $307.38

AAJ66985MCR

 
alfa aesar™
J66985MCR
1mg Each for $103.22
Description & Specifications

Specifications

Formula Weight 502.51
Chemical Name or Material N-Acetyl-Ile-Glu-Thr-Asp-al
CAS 191338-86-0
MDL Number MFCD01318858
Synonym Acetyl-IETD-CHO; Caspase-8 Inhibitor I
Physical Form Lyophilized Powder
Color White to off-white
Quantity 5mg
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C