Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Ile-Glu-Pro-Asp-al

Chemical Identifiers

Chemical Name or Material N-Acetyl-Ile-Glu-Pro-Asp-al
Formula Weight 498.53
MDL Number MFCD02259689
Synonym Ac-IEPD-CHO; Caspase-8 Inhibitor
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66294LB0

 
alfa aesar™
J66294LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66294MCR

 
alfa aesar™
J66294MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 498.53
Chemical Name or Material N-Acetyl-Ile-Glu-Pro-Asp-al
Synonym Ac-IEPD-CHO; Caspase-8 Inhibitor
Physical Form Lyophilized Powder
MDL Number MFCD02259689
Storage Note 1 -30° to -10°C
Quantity 5mg
Color White to off-white
Safety and Handling

-30° to -10°C