Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Glu-Glu-Val-Val-Pro-Cys-p-nitroanilide

Chemical Identifiers

Chemical Name or Material N-Acetyl-Glu-Glu-Val-Val-Pro-Cys-p-nitroanilide
Formula Weight 836.91
MDL Number MFCD07366698
Synonym Ac-Glu-Glu-Val-Val-Pro-Cys-pNA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66897EXD

 
alfa aesar™
J66897EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66897MCR

 
alfa aesar™
J66897MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 836.91
Chemical Name or Material N-Acetyl-Glu-Glu-Val-Val-Pro-Cys-p-nitroanilide
Physical Form Solid
Synonym Ac-Glu-Glu-Val-Val-Pro-Cys-pNA
MDL Number MFCD07366698
Color White to off-white
Quantity 0.5mg
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C