Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-DL-valine, 99+%

$33.76 - $86.19

Chemical Identifiers

Chemical Name or Material N-Acetyl-DL-valine
CAS 3067-19-4
Molecular Formula C7H13NO3
Formula Weight 159.19
MDL Number MFCD00066065
Beilstein 1723835
Synonym Ac-DL-Val-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1624406

  SDS
alfa aesar™
L1624406
5g Each for $33.76

AAL1624414

  SDS
alfa aesar™
L1624414
25g Each for $86.19
Description & Specifications

Specifications

Formula Weight 159.19
CAS 3067-19-4
Chemical Name or Material N-Acetyl-DL-valine
Assay ≥99%
Molecular Formula C7H13NO3
MDL Number MFCD00066065
Beilstein 1723835
Synonym Ac-DL-Val-OH
Melting Point 146° to 149°C
Quantity 5g