missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-acetyl-DL-tryptophan, ∽99%, MP Biomedicals™

N-acetyl-DL-tryptophan, is used as stabilizer in the human blood-derived therapeutic products normal serum albumin and plasma protein fraction

Supplier:  MP Biomedicals Inc 0210014291

 View more versions of this product

Catalog No. ICN10014291


Explore available promotions

Description

Description

Specifications

Specifications

N-acetyl-DL-tryptophan
White
C13H14N2O3
n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
246.27
CHEBI:70976
1 kg
87-32-1
∽99%
MFCD00005644
DZTHIGRZJZPRDV-UHFFFAOYSA-N
2-acetamido-3-(1H-indol-3-yl)propanoic acid
2002
246.3
Powder

Chemical Identifiers

87-32-1
246.27
DZTHIGRZJZPRDV-UHFFFAOYSA-N
2002
2-acetamido-3-(1H-indol-3-yl)propanoic acid
C13H14N2O3
MFCD00005644
n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
CHEBI:70976
CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
Safety and Handling

Safety and Handling

EINECSNumber : 201-739-3

Recommended Storage : Store at +4°C, Store Desiccated.

SDS
Documents

Documents

Promotions

Promotions