Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-DL-phenylglycine, 98%

Chemical Identifiers

Chemical Name or Material N-Acetyl-DL-phenylglycine
CAS 15962-46-6
Formula Weight 193.2
MDL Number MFCD00055924
Synonym Ac-DL-Phg-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6613606

 
alfa aesar™
H6613606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6613622

 
alfa aesar™
H6613622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6613614

 
alfa aesar™
H6613614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.2
Chemical Name or Material N-Acetyl-DL-phenylglycine
CAS 15962-46-6
Physical Form Powder
MDL Number MFCD00055924
Assay 98%
Synonym Ac-DL-Phg-OH
Storage Note 1 -30° to -10°C
Color White
Quantity 5g
Safety and Handling

-30° to -10°C