Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-DL-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material N-Acetyl-DL-phenylalanine
CAS 2901-75-9
Formula Weight 207.23
MDL Number MFCD00063157
Synonym Ac-DL-Phe-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6312514

 
alfa aesar™
H6312514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6312522

 
alfa aesar™
H6312522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.23
Chemical Name or Material N-Acetyl-DL-phenylalanine
Physical Form Powder
Assay 95%
MDL Number MFCD00063157
CAS 2901-75-9
Synonym Ac-DL-Phe-OH
Color Off-white
Quantity 25g
Storage Note 1 -30° to -10°C
RTECS Number SQ7321132
Safety and Handling

-30° to -10°C