Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-DL-norvaline, 98%

$124.68 - $1,714.76

Chemical Identifiers

Chemical Name or Material N-Acetyl-DL-norvaline
CAS 7682-15-7
Molecular Formula C7H13NO3
Formula Weight 159.19
MDL Number MFCD00037269
Synonym 2-Acetamidopentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2538903

 
alfa aesar™
B2538903
1g Each for $124.68

AAB2538906

 
alfa aesar™
B2538906
5g Each for $438.69

AAB2538914

 
alfa aesar™
B2538914
25g Each for $1,714.76
Description & Specifications

Specifications

Formula Weight 159.19
Chemical Name or Material N-Acetyl-DL-norvaline
Assay 98%
MDL Number MFCD00037269
CAS 7682-15-7
Molecular Formula C7H13NO3
Synonym 2-Acetamidopentanoic acid
Quantity 1g