Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-DL-histidine hydrate, 98%

Chemical Identifiers

Chemical Name or Material N-Acetyl-DL-histidine hydrate
CAS 213178-97-3
Formula Weight 197.19 (Anhydrous)
MDL Number MFCD00150969
Synonym Ac-DL-His-OH.H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6685403

 
alfa aesar™
H6685403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6685406

 
alfa aesar™
H6685406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.19 (Anhydrous)
Chemical Name or Material N-Acetyl-DL-histidine hydrate
Physical Form Solid
Assay 98%
MDL Number MFCD00150969
CAS 213178-97-3
Quantity 1g
Synonym Ac-DL-His-OH.H2O