Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-DL-4-fluorophenylalanine, 98%

$83.93 - $83.93

Chemical Identifiers

Chemical Name or Material N-Acetyl-DL-4-fluorophenylalanine
CAS 17481-06-0
Molecular Formula C11H12FNO3
Formula Weight 225.22
MDL Number MFCD00017983
Beilstein 3206105
Synonym Ac-DL-Phe(4-F)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0847406

  SDS
alfa aesar™
L0847406
5g Each for $83.93
Description & Specifications

Specifications

Formula Weight 225.22
Chemical Name or Material N-Acetyl-DL-4-fluorophenylalanine
MDL Number MFCD00017983
Molecular Formula C11H12FNO3
CAS 17481-06-0
Assay 98%
Recommended Storage 2° to 8°C
Melting Point 150° to 153°C
Beilstein 3206105
Synonym Ac-DL-Phe(4-F)-OH
TSCA TSCA
Quantity 5g
Safety and Handling

2° to 8°C