Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-D-valine, 98%

Chemical Identifiers

Chemical Name or Material N-Acetyl-D-valine
CAS 17916-88-0
Formula Weight 159.19
MDL Number MFCD03939895
Synonym Ac-D-val-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6610306

 
alfa aesar™
H6610306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6610314

 
alfa aesar™
H6610314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6610322

 
alfa aesar™
H6610322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 159.19
Chemical Name or Material N-Acetyl-D-valine
MDL Number MFCD03939895
CAS 17916-88-0
Assay 98%
Synonym Ac-D-val-OH
Quantity 5g
Physical Form Solid
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C