missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-D-tryptophan 98.0+%, TCI America™

$34.55 - $95.78

Chemical Identifiers

CAS 2280-01-5
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00065021
InChI Key DZTHIGRZJZPRDV-GFCCVEGCSA-N
Synonym Ac-D-Trp-OH
PubChem CID 439917
ChEBI CHEBI:16734
IUPAC Name (2R)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A01191G View Documents Tci America
A01191G
1g Each for $34.55
 
A01195G View Documents Tci America
A01195G
5g Each for $95.78
 
Specifications

Chemical Identifiers

2280-01-5
246.27
DZTHIGRZJZPRDV-GFCCVEGCSA-N
439917
(2R)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
C13H14N2O3
MFCD00065021
Ac-D-Trp-OH
CHEBI:16734
CC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)O

Specifications

White
1g
C13H14N2O3
Ac-D-Trp-OH
CC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)O
246.27
CHEBI:16734
Crystal-Powder at 20°C
N-Acetyl-D-tryptophan
186°C
2280-01-5
MFCD00065021
DZTHIGRZJZPRDV-GFCCVEGCSA-N
(2R)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
439917
246.27
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1601

TSCA : No