Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-D-proline, 98%

Chemical Identifiers

Chemical Name or Material N-Acetyl-D-proline
CAS 59785-68-1
Formula Weight 157.17
MDL Number MFCD00080841
Synonym Ac-D-Pro-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6652406

 
alfa aesar™
H6652406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6652414

 
alfa aesar™
H6652414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 157.17
MDL Number MFCD00080841
Chemical Name or Material N-Acetyl-D-proline
Assay 98%
CAS 59785-68-1
Storage Note 1 -30° to -10°C
Physical Form Solid
Synonym Ac-D-Pro-OH
Quantity 5g
Safety and Handling

-30° to -10°C