Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material N-Acetyl-D-phenylalanine
CAS 10172-89-1
Formula Weight 207.23
MDL Number MFCD00002664
Beilstein 2213852
Synonym Ac-D-Phe-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6338206

 
alfa aesar™
H6338206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6338214

 
alfa aesar™
H6338214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6338222

 
alfa aesar™
H6338222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.23
Melting Point 165° to 169°C
Chemical Name or Material N-Acetyl-D-phenylalanine
Physical Form Solid
Assay 95%
MDL Number MFCD00002664
CAS 10172-89-1
Beilstein 2213852
Synonym Ac-D-Phe-OH
Color White
Quantity 5g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C