Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-D-methionine, 98%

Chemical Identifiers

Chemical Name or Material N-Acetyl-D-methionine
CAS 1509-92-8
Formula Weight 191.25
MDL Number MFCD00069962
Beilstein 1725553
Synonym Ac-D-Met-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6680806

 
alfa aesar™
H6680806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6680814

 
alfa aesar™
H6680814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6680822

 
alfa aesar™
H6680822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.25
Melting Point 102° to 104°C
Chemical Name or Material N-Acetyl-D-methionine
Physical Form Crystalline
CAS 1509-92-8
Synonym Ac-D-Met-OH
Assay 98%
MDL Number MFCD00069962
Beilstein 1725553
Color White
Quantity 5g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C